The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133467.2 | NP_597724.1 | 1 - 1 | 275 | 90 |
GenBank mRNA | BC035496.2 | - | 1 - 1 | 277 | 90 |
BC052559.1 | - | 1 - 1 | 275 | 90 |