Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 4 - 5 4120 94
NM_014866.1 NP_055681.1 5 - 6 4110 94
XM_005266134.2 XP_005266191.1 4 - 5 3973 94
XM_011519246.2 XP_011517548.1 5 - 6 4119 94
XM_011519247.1 XP_011517549.1 4 - 5 3976 94
XM_011519248.2 XP_011517550.1 4 - 5 3950 94
XM_011519249.1 XP_011517551.1 4 - 5 3973 94
XM_011519250.2 XP_011517552.1 4 - 5 3973 94
XM_011519251.1 XP_011517553.1 4 - 5 3973 94
XM_011519252.2 XP_011517554.1 4 - 5 3973 94
XM_011519253.1 XP_011517555.1 4 - 5 3973 94
XM_011519254.1 XP_011517556.1 4 - 5 3973 94
XM_011519255.2 XP_011517557.1 4 - 5 3973 94
XM_011519256.1 XP_011517558.1 4 - 5 3973 94
XM_011519257.1 XP_011517559.1 4 - 5 3973 94
XM_011519258.1 XP_011517560.1 4 - 5 3973 94
XM_011519259.1 XP_011517561.1 4 - 5 3973 94
XM_011519260.2 XP_011517562.1 4 - 5 3957 94
XM_011519261.1 XP_011517563.1 4 - 5 3973 94
XM_011519262.1 XP_011517564.1 5 - 6 3262 94
XM_011519263.2 XP_011517565.1 5 - 6 3262 94
XM_011519264.2 XP_011517566.1 4 - 5 3973 94
XM_017015363.1 XP_016870852.1 4 - 5 3973 94
XM_017015364.1 XP_016870853.1 4 - 5 3973 94
GenBank mRNA AB002308.4 - 5 - 6 4110 94
AB593115.1 - 4 - 5 4120 94
BC125018.1 - 3 - 4 2682 94
BC125019.1 - 4 - 5 1971 94
DQ903855.1 - 3 - 4 3267 94

View Full Product Details