Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUN2
Entrez Gene ID
25777
Gene Name
Sad1 and UNC84 domain containing 2
Gene Aliases
UNC84B
Chromosome Location
Chr.22: 38734714 - 38756019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199579.1 NP_001186508.1 8 - 9 1172 86
NM_001199580.1 NP_001186509.1 9 - 10 1108 86
NM_015374.2 NP_056189.1 8 - 9 1109 86
XM_011530104.2 XP_011528406.1 7 - 8 966 86
XM_011530105.2 XP_011528407.1 8 - 9 833 86
XM_017028748.1 XP_016884237.1 8 - 9 1104 86
GenBank mRNA AB014568.1 - 8 - 9 891 86
AK295787.1 - 8 - 9 833 86
AK304188.1 - 9 - 10 968 86
AY682988.1 - 8 - 9 890 86
BC094797.1 - 8 - 9 1129 86
BC111549.1 - 7 - 8 838 86
BC111717.1 - 7 - 8 838 86
BX537962.1 - 9 - 10 1100 86
CR456474.1 - 7 - 8 836 86

View Full Product Details