Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUN2
Entrez Gene ID
25777
Gene Name
Sad1 and UNC84 domain containing 2
Gene Aliases
UNC84B
Chromosome Location
Chr.22: 38734714 - 38756019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199579.1 NP_001186508.1 13 - 14 1933 61
NM_001199580.1 NP_001186509.1 14 - 15 1869 61
NM_015374.2 NP_056189.1 13 - 14 1870 61
XM_011530104.2 XP_011528406.1 12 - 13 1727 61
XM_011530105.2 XP_011528407.1 13 - 14 1594 61
XM_017028748.1 XP_016884237.1 13 - 14 1865 61
GenBank mRNA AB014568.1 - 13 - 14 1652 61
AF202723.1 - 5 - 6 745 61
AK295787.1 - 13 - 14 1594 61
AK304188.1 - 14 - 15 1729 61
AY682988.1 - 13 - 14 1651 61
BC030684.2 - 1 - 2 75 61
BC069253.1 - 5 - 6 751 61
BC094797.1 - 13 - 14 1890 61
BC111549.1 - 12 - 13 1599 61
BC111717.1 - 12 - 13 1599 61
BX537962.1 - 14 - 15 1861 61
CR456474.1 - 12 - 13 1597 61

View Full Product Details