The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015315.4 | NP_056130.2 | 3 - 4 | 354 | 59 |
XM_005268404.3 | XP_005268461.1 | 3 - 4 | 744 | 59 | |
XM_005268406.3 | XP_005268463.1 | 3 - 4 | 744 | 59 | |
XM_005268407.3 | XP_005268464.1 | 3 - 4 | 500 | 59 | |
XM_005268408.3 | XP_005268465.1 | 3 - 4 | 237 | 59 | |
XM_006714775.2 | XP_006714838.1 | 3 - 4 | 153 | 59 | |
XM_006714776.3 | XP_006714839.1 | 3 - 4 | 258 | 59 | |
XM_011537614.1 | XP_011535916.1 | 3 - 4 | 744 | 59 | |
XM_011537615.1 | XP_011535917.1 | 3 - 4 | 744 | 59 | |
XM_011537616.1 | XP_011535918.1 | 3 - 4 | 252 | 59 | |
XM_011537617.1 | XP_011535919.1 | 3 - 4 | 171 | 59 | |
XM_011537618.2 | XP_011535920.1 | 3 - 4 | 305 | 59 | |
XM_017009283.1 | XP_016864772.1 | 3 - 4 | 500 | 59 | |
XM_017009284.1 | XP_016864773.1 | 3 - 4 | 500 | 59 | |
GenBank mRNA | AB018274.1 | - | 4 - 5 | 561 | 59 |
AK091465.1 | - | 3 - 4 | 744 | 59 | |
AK307554.1 | - | 3 - 4 | 213 | 59 | |
BC001460.2 | - | 3 - 4 | 354 | 59 |