The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304331.1 | NP_001291260.1 | 25 - 26 | 3560 | 63 |
NM_001304332.1 | NP_001291261.1 | 24 - 25 | 3533 | 63 | |
XM_006711586.2 | XP_006711649.1 | 25 - 26 | 3590 | 63 | |
XM_011510070.2 | XP_011508372.1 | 13 - 14 | 1619 | 63 | |
XM_017002606.1 | XP_016858095.1 | 24 - 25 | 3563 | 63 | |
XM_017002607.1 | XP_016858096.1 | 24 - 25 | 3455 | 63 | |
XM_017002608.1 | XP_016858097.1 | 24 - 25 | 3563 | 63 | |
XM_017002609.1 | XP_016858098.1 | 24 - 25 | 3455 | 63 | |
GenBank mRNA | AB020704.1 | - | 7 - 8 | 689 | 63 |
AF034801.1 | - | 9 - 10 | 1084 | 63 | |
AK023365.1 | - | 6 - 7 | 730 | 63 | |
AK124535.1 | - | 1 - 2 | 1047 | 63 | |
AK126624.1 | - | 12 - 13 | 2128 | 63 | |
AK295757.1 | - | 19 - 20 | 2288 | 63 | |
BC132923.1 | - | 12 - 13 | 2070 | 63 | |
BC132925.1 | - | 12 - 13 | 2070 | 63 | |
BC144262.1 | - | 11 - 12 | 2043 | 63 | |
EF428334.1 | - | 30 - 31 | 3427 | 63 |