The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304331.1 | NP_001291260.1 | 26 - 27 | 3657 | 64 |
NM_001304332.1 | NP_001291261.1 | 25 - 26 | 3630 | 64 | |
XM_006711586.2 | XP_006711649.1 | 26 - 27 | 3687 | 64 | |
XM_011510070.2 | XP_011508372.1 | 14 - 15 | 1716 | 64 | |
XM_017002606.1 | XP_016858095.1 | 25 - 26 | 3660 | 64 | |
XM_017002607.1 | XP_016858096.1 | 25 - 26 | 3552 | 64 | |
XM_017002608.1 | XP_016858097.1 | 25 - 26 | 3660 | 64 | |
XM_017002609.1 | XP_016858098.1 | 25 - 26 | 3552 | 64 | |
GenBank mRNA | AB020704.1 | - | 8 - 9 | 786 | 64 |
AF034801.1 | - | 10 - 11 | 1181 | 64 | |
AK023365.1 | - | 7 - 8 | 827 | 64 | |
AK124535.1 | - | 2 - 3 | 1144 | 64 | |
AK126624.1 | - | 13 - 14 | 2225 | 64 | |
AK295757.1 | - | 20 - 21 | 2385 | 64 | |
BC132923.1 | - | 13 - 14 | 2167 | 64 | |
BC132925.1 | - | 13 - 14 | 2167 | 64 | |
BC144262.1 | - | 12 - 13 | 2140 | 64 | |
EF428334.1 | - | 31 - 32 | 3524 | 64 |