The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308330.1 | NP_001295259.1 | 12 - 13 | 1324 | 152 |
NM_014980.2 | NP_055795.1 | 12 - 13 | 1324 | 152 | |
XM_006713825.3 | XP_006713888.1 | 12 - 13 | 1494 | 152 | |
XM_011513331.2 | XP_011511633.1 | 12 - 13 | 1321 | 152 | |
XM_011513333.2 | XP_011511635.1 | 12 - 13 | 1321 | 152 | |
XM_017007530.1 | XP_016863019.1 | 12 - 13 | 1255 | 152 | |
XM_017007531.1 | XP_016863020.1 | 12 - 13 | 1309 | 152 | |
XM_017007532.1 | XP_016863021.1 | 12 - 13 | 1258 | 152 | |
XM_017007533.1 | XP_016863022.1 | 9 - 10 | 912 | 152 | |
XM_017007534.1 | XP_016863023.1 | 12 - 13 | 1321 | 152 | |
XM_017007535.1 | XP_016863024.1 | 12 - 13 | 1512 | 152 | |
GenBank mRNA | AB023223.1 | - | 12 - 13 | 1291 | 152 |
AK296544.1 | - | 12 - 13 | 1322 | 152 |