Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
Chromosome Location
Chr.6: 24171755 - 24383292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195610.1 NP_001182539.1 3 - 4 561 117
NM_016356.4 NP_057440.2 2 - 3 654 117
GenBank mRNA AB032980.2 - 2 - 3 352 117
AF181721.1 - 2 - 3 654 117
AK027036.1 - 2 - 3 470 117
BC050704.1 - 3 - 4 561 117
BM923267.1 - 3 - 4 574 117

View Full Product Details