Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC2
Entrez Gene ID
51473
Gene Name
doublecortin domain containing 2
Gene Aliases
DCDC2A, DFNB66, NPHP19, RU2, RU2S
Chromosome Location
Chr.6: 24171755 - 24383292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195610.1 NP_001182539.1 9 - 10 1231 95
NM_016356.4 NP_057440.2 8 - 9 1324 95
GenBank mRNA AB032980.2 - 8 - 9 1022 95
AF181721.1 - 8 - 9 1324 95
AK027036.1 - 8 - 9 1140 95
AL133043.1 - 1 - 2 303 95
BC050704.1 - 9 - 10 1231 95

View Full Product Details