The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020718.3 | NP_065769.3 | 12 - 13 | 1948 | 73 |
XM_017023497.1 | XP_016878986.1 | 12 - 13 | 3474 | 73 | |
XM_017023498.1 | XP_016878987.1 | 11 - 12 | 1261 | 73 | |
XM_017023499.1 | XP_016878988.1 | 9 - 10 | 1065 | 73 | |
XM_017023500.1 | XP_016878989.1 | 12 - 13 | 1979 | 73 | |
GenBank mRNA | AB033029.1 | - | 1 - 2 | 73 | 73 |
AJ586135.2 | - | 12 - 13 | 1948 | 73 | |
AK126752.1 | - | 12 - 13 | 1632 | 73 | |
BX648357.1 | - | 3 - 4 | 336 | 73 |