Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
Chromosome Location
Chr.7: 105604774 - 105877232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020725.1 NP_065776.1 11 - 12 2574 81
XM_011515935.1 XP_011514237.1 13 - 14 2963 81
XM_011515939.1 XP_011514241.1 13 - 14 2687 81
XM_011515940.1 XP_011514242.1 12 - 13 2657 81
GenBank mRNA AB033044.1 - 11 - 12 2587 81

View Full Product Details