Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
Chromosome Location
Chr.9: 72114595 - 72256045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242505.2 NP_001229434.1 10 - 11 1200 130
NM_001242506.2 NP_001229435.1 11 - 12 1241 130
NM_001242507.2 NP_001229436.1 10 - 11 1107 130
NM_004293.4 NP_004284.1 10 - 11 1200 130
XM_005252317.1 XP_005252374.1 10 - 11 1198 130
XM_011519213.1 XP_011517515.1 11 - 12 1312 130
XM_011519214.1 XP_011517516.1 11 - 12 1312 130
XM_011519215.1 XP_011517517.1 11 - 12 1312 130
XM_011519216.1 XP_011517518.1 11 - 12 1312 130
XM_011519217.1 XP_011517519.1 11 - 12 1312 130
XM_011519218.2 XP_011517520.1 11 - 12 1197 130
XM_011519219.1 XP_011517521.1 11 - 12 1310 130
XM_011519220.1 XP_011517522.1 10 - 11 1196 130
XM_011519221.1 XP_011517523.1 10 - 11 1107 130
XM_011519222.1 XP_011517524.1 11 - 12 1312 130
XM_011519223.2 XP_011517525.1 8 - 9 677 130
XM_017015333.1 XP_016870822.1 11 - 12 1312 130
XM_017015334.1 XP_016870823.1 11 - 12 1312 130
XM_017015335.1 XP_016870824.1 11 - 12 1312 130
XM_017015336.1 XP_016870825.1 10 - 11 1198 130
XM_017015337.1 XP_016870826.1 10 - 11 1198 130
XM_017015338.1 XP_016870827.1 12 - 13 1268 130
XM_017015339.1 XP_016870828.1 10 - 11 1196 130
XM_017015340.1 XP_016870829.1 8 - 9 716 130
GenBank mRNA AB033084.1 - 10 - 11 1067 130
AF019638.1 - 10 - 11 1076 130
AF095286.1 - 10 - 11 1084 130
AK295716.1 - 3 - 4 242 130
AK300418.1 - 10 - 11 1107 130
AK315988.1 - 11 - 12 1019 130
BC053584.1 - 10 - 11 1174 130

View Full Product Details