The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017553.2 | NP_060023.1 | 7 - 8 | 1191 | 72 |
NR_104038.1 | 7 - 8 | 1191 | 72 | ||
XM_011521685.2 | XP_011519987.1 | 7 - 8 | 2026 | 72 | |
GenBank mRNA | AB033085.1 | - | 7 - 8 | 1087 | 72 |
AK290371.1 | - | 6 - 7 | 929 | 72 | |
BC144681.1 | - | 7 - 8 | 1077 | 72 | |
BC146785.1 | - | 7 - 8 | 1099 | 72 |