Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LST1
Entrez Gene ID
7940
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144, D6S49E, LST-1
Chromosome Location
Chr.6: 31586179 - 31588909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205838.2 NP_995310.2 1 - 2 133 73
NM_205839.2 NP_995311.2 1 - 2 133 73
NR_029461.1 1 - 2 133 73
NR_029462.1 1 - 2 133 73
XM_006715206.3 XP_006715269.1 1 - 2 125 73
XM_006715209.3 XP_006715272.1 1 - 2 121 73
XM_006715210.3 XP_006715273.1 1 - 2 127 73
GenBank mRNA AF000425.1 - 1 - 2 111 73
AJ705986.1 - 1 - 2 70 73
AV711621.1 - 1 - 2 97 73
AV715235.1 - 1 - 2 188 73
AW925228.1 - 1 - 2 133 73
BX647293.1 - 1 - 2 118 73

View Full Product Details