The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144994.1 | NP_001138466.1 | 2 - 3 | 748 | 54 |
GenBank mRNA | AF007149.1 | - | 1 - 2 | 153 | 54 |
DA141613.1 | - | 2 - 3 | 198 | 54 |