The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075098.3 | NP_001068566.1 | 5 - 6 | 780 | 62 |
NM_005943.5 | NP_005934.2 | 4 - 5 | 784 | 62 | |
NR_033233.1 | 4 - 5 | 653 | 62 | ||
XM_011514632.2 | XP_011512934.1 | 5 - 6 | 691 | 62 | |
XM_011514633.2 | XP_011512935.1 | 5 - 6 | 691 | 62 | |
XM_011514634.2 | XP_011512936.1 | 4 - 5 | 588 | 62 | |
XM_017010888.1 | XP_016866377.1 | 4 - 5 | 598 | 62 | |
GenBank mRNA | AF034374.1 | - | 5 - 6 | 663 | 62 |
AJ224328.1 | - | 4 - 5 | 781 | 62 | |
AK056740.1 | - | 4 - 5 | 599 | 62 | |
AK091306.1 | - | 4 - 5 | 512 | 62 | |
AK300300.1 | - | 5 - 6 | 653 | 62 | |
BC036839.1 | - | 5 - 6 | 653 | 62 | |
CX787499.1 | - | 1 - 2 | 61 | 62 |