Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R35
Entrez Gene ID
221908
Gene Name
protein phosphatase 1 regulatory subunit 35
Gene Aliases
C7orf47
Chromosome Location
Chr.7: 100435282 - 100436615 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145030.2 NP_659467.1 3 - 4 101
XM_011515914.1 XP_011514216.1 3 - 4 101
GenBank mRNA BC026269.1 - 3 - 4 101

View Full Product Details