Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
Chromosome Location
Chr.14: 34561409 - 34630160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021249.3 NP_067072.3 3 - 4 421 89
NM_152233.2 NP_689419.2 4 - 5 316 89
GenBank mRNA AF121856.1 - 4 - 5 307 89
AK292250.1 - 4 - 5 299 89
AK296213.1 - 4 - 5 310 89
AK308464.1 - 4 - 5 310 89
BC001798.1 - 3 - 4 421 89
BG722853.1 - 4 - 5 342 89
U83194.1 - 4 - 5 284 89

View Full Product Details