The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082967.2 | NP_001076436.1 | 2 - 3 | 395 | 114 |
NM_015381.6 | NP_056196.2 | 2 - 3 | 416 | 114 | |
GenBank mRNA | AF131851.1 | - | 2 - 3 | 361 | 114 |
AK123443.1 | - | 3 - 4 | 355 | 114 | |
AK125256.1 | - | 2 - 3 | 416 | 114 | |
AY325118.1 | - | 2 - 3 | 390 | 114 | |
AY358847.1 | - | 3 - 4 | 291 | 114 | |
BC039396.1 | - | 2 - 3 | 374 | 114 |