The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173513.1 | NP_001166984.1 | 6 - 7 | 848 | 78 |
NM_052883.1 | NP_443115.1 | 6 - 7 | 848 | 78 | |
GenBank mRNA | AF133519.1 | - | 6 - 7 | 516 | 78 |
AF171055.1 | - | 6 - 7 | 522 | 78 | |
AK299502.1 | - | 6 - 7 | 825 | 78 | |
BC050032.1 | - | 6 - 7 | 835 | 78 |