Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
Chromosome Location
Chr.9: 37509920 - 37576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012166.2 NP_036298.2 2 - 3 626 78
XM_005251439.4 XP_005251496.1 2 - 3 715 78
XM_017014618.1 XP_016870107.1 2 - 3 832 78
XM_017014619.1 XP_016870108.1 2 - 3 943 78
GenBank mRNA AB209610.1 - 1 - 2 214 78
AF176705.1 - 2 - 3 625 78
BC125125.1 - 2 - 3 705 78
BC140785.1 - 1 - 2 597 78
BC171785.1 - 1 - 2 597 78

View Full Product Details