The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020795.3 | NP_065846.1 | 2 - 3 | 578 | 77 |
XM_017024897.1 | XP_016880386.1 | 2 - 3 | 167 | 77 | |
GenBank mRNA | AF376802.1 | - | 2 - 3 | 578 | 77 |