The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204197.1 | NP_001191126.1 | 8 - 9 | 956 | 71 |
NM_015678.4 | NP_056493.3 | 48 - 49 | 7986 | 71 | |
XM_005266346.4 | XP_005266403.1 | 48 - 49 | 7986 | 71 | |
XM_005266347.4 | XP_005266404.2 | 48 - 49 | 7986 | 71 | |
XM_005266348.4 | XP_005266405.2 | 47 - 48 | 7977 | 71 | |
XM_006719803.3 | XP_006719866.1 | 48 - 49 | 7986 | 71 | |
XM_006719805.3 | XP_006719868.1 | 47 - 48 | 7977 | 71 | |
XM_006719806.3 | XP_006719869.1 | 47 - 48 | 7977 | 71 | |
XM_011535046.2 | XP_011533348.1 | 40 - 41 | 6117 | 71 | |
XM_017020544.1 | XP_016876033.1 | 48 - 49 | 7968 | 71 | |
XM_017020545.1 | XP_016876034.1 | 48 - 49 | 7968 | 71 | |
XM_017020546.1 | XP_016876035.1 | 48 - 49 | 7274 | 71 | |
GenBank mRNA | AB046764.1 | - | 15 - 16 | 1698 | 71 |
AF072371.1 | - | 2 - 3 | 173 | 71 | |
AF467288.1 | - | 49 - 50 | 7658 | 71 | |
AK091469.1 | - | 19 - 20 | 2145 | 71 | |
AK122888.1 | - | 18 - 19 | 2499 | 71 | |
AK127791.1 | - | 25 - 26 | 3337 | 71 | |
AK294737.1 | - | 8 - 9 | 956 | 71 | |
AK297773.1 | - | 9 - 10 | 1054 | 71 | |
AK315939.1 | - | 8 - 9 | 953 | 71 | |
AL137748.1 | - | 2 - 3 | 598 | 71 | |
BX640760.1 | - | 24 - 25 | 3316 | 71 |