Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NBEA
Entrez Gene ID
26960
Gene Name
neurobeachin
Gene Aliases
BCL8B, LYST2
Chromosome Location
Chr.13: 34942287 - 35672737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204197.1 NP_001191126.1 12 - 13 1476 72
NM_015678.4 NP_056493.3 52 - 53 8506 72
XM_005266346.4 XP_005266403.1 52 - 53 8491 72
XM_005266347.4 XP_005266404.2 53 - 54 8569 72
XM_005266348.4 XP_005266405.2 52 - 53 8560 72
XM_006719803.3 XP_006719866.1 53 - 54 8554 72
XM_006719805.3 XP_006719868.1 51 - 52 8482 72
XM_006719806.3 XP_006719869.1 51 - 52 8497 72
XM_011535046.2 XP_011533348.1 45 - 46 6700 72
XM_017020544.1 XP_016876033.1 53 - 54 8551 72
XM_017020545.1 XP_016876034.1 52 - 53 8488 72
XM_017020546.1 XP_016876035.1 53 - 54 7857 72
GenBank mRNA AB046764.1 - 19 - 20 2218 72
AF072371.1 - 6 - 7 693 72
AF467288.1 - 53 - 54 8178 72
AK091469.1 - 23 - 24 2665 72
AK122888.1 - 22 - 23 3019 72
AK127791.1 - 29 - 30 3857 72
AK294737.1 - 12 - 13 1476 72
AL137748.1 - 7 - 8 1181 72
BX640760.1 - 28 - 29 3836 72

View Full Product Details