The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098802.1 | NP_001092272.1 | 3 - 4 | 771 | 123 |
NM_032171.1 | NP_115547.1 | 3 - 4 | 771 | 123 | |
XM_005252263.4 | XP_005252320.1 | 3 - 4 | 784 | 123 | |
XM_005252265.2 | XP_005252322.1 | 3 - 4 | 784 | 123 | |
XM_005252266.4 | XP_005252323.1 | 3 - 4 | 778 | 123 | |
XM_017015192.1 | XP_016870681.1 | 3 - 4 | 784 | 123 | |
XM_017015193.1 | XP_016870682.1 | 3 - 4 | 784 | 123 | |
XM_017015194.1 | XP_016870683.1 | 3 - 4 | 784 | 123 | |
XM_017015195.1 | XP_016870684.1 | 3 - 4 | 784 | 123 | |
XM_017015196.1 | XP_016870685.1 | 3 - 4 | 778 | 123 | |
GenBank mRNA | AK022705.1 | - | 3 - 4 | 751 | 123 |
AK303457.1 | - | 3 - 4 | 821 | 123 | |
BC058931.1 | - | 3 - 4 | 684 | 123 | |
BC128058.1 | - | 3 - 4 | 639 | 123 |