The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021228.2 | NP_067051.2 | 7 - 8 | 3443 | 96 |
XM_005259122.4 | XP_005259179.1 | 7 - 8 | 3726 | 96 | |
XM_011527194.2 | XP_011525496.1 | 7 - 8 | 3368 | 96 | |
XM_017027083.1 | XP_016882572.1 | 4 - 5 | 3170 | 96 | |
GenBank mRNA | AK024444.1 | - | 7 - 8 | 3443 | 96 |
BC018398.1 | - | 1 - 2 | 834 | 96 | |
BC023628.2 | - | 1 - 2 | 1210 | 96 | |
BC053992.1 | - | 6 - 7 | 3330 | 96 |