The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022370.3 | NP_071765.2 | 3 - 4 | 810 | 78 |
XM_017018122.1 | XP_016873611.1 | 2 - 3 | 575 | 78 | |
GenBank mRNA | AK291045.1 | - | 3 - 4 | 744 | 78 |
AY509035.1 | - | 3 - 4 | 833 | 78 |