Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PM20D1
Entrez Gene ID
148811
Gene Name
peptidase M20 domain containing 1
Gene Aliases
Cps1
Chromosome Location
Chr.1: 205813811 - 205850148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152491.4 NP_689704.4 10 - 11 1196 82
NR_135186.1 9 - 10 1134 82
XM_017000396.1 XP_016855885.1 10 - 11 1190 82
GenBank mRNA AK057131.1 - 9 - 10 1118 82
AK290786.1 - 10 - 11 1180 82
BC039170.1 - 10 - 11 1165 82
BC063477.1 - 10 - 11 1165 82

View Full Product Details