The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033054.2 | NP_149043.2 | 7 - 8 | 993 | 56 |
XM_017012503.1 | XP_016867992.1 | 6 - 7 | 752 | 56 | |
XM_017012504.1 | XP_016867993.1 | 7 - 8 | 988 | 56 | |
GenBank mRNA | AB290179.1 | - | 7 - 8 | 953 | 56 |
AF380932.1 | - | 6 - 7 | 753 | 56 | |
AF380933.1 | - | 6 - 7 | 753 | 56 | |
AK074050.1 | - | 6 - 7 | 3836 | 56 | |
AK074135.1 | - | 3 - 4 | 273 | 56 | |
AK097912.1 | - | 7 - 8 | 984 | 56 | |
AK131089.1 | - | 2 - 3 | 3110 | 56 | |
AL832302.1 | - | 6 - 7 | 748 | 56 | |
BC113544.1 | - | 7 - 8 | 940 | 56 | |
BC117241.1 | - | 7 - 8 | 940 | 56 |