The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002188.3 | 3 - 4 | 343 | 74 | |
GenBank mRNA | AK295259.1 | - | 2 - 3 | 253 | 74 |
BC030240.1 | - | 3 - 4 | 222 | 74 | |
BC092482.1 | - | 2 - 3 | 253 | 74 | |
KJ690774.1 | - | 2 - 3 | 171 | 74 | |
KJ690775.1 | - | 2 - 3 | 171 | 74 | |
KJ690776.1 | - | 2 - 3 | 171 | 74 |