Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location
Chr.9: 98732009 - 98796542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173551.4 NP_775822.3 13 - 14 2443 112
XM_005251793.4 XP_005251850.1 13 - 14 2433 112
XM_005251794.4 XP_005251851.1 13 - 14 2430 112
XM_006716998.3 XP_006717061.1 13 - 14 2433 112
XM_006716999.2 XP_006717062.1 13 - 14 3518 112
XM_011518357.2 XP_011516659.1 13 - 14 4515 112
XM_017014445.1 XP_016869934.1 13 - 14 2430 112
XM_017014446.1 XP_016869935.1 12 - 13 4394 112
XM_017014447.1 XP_016869936.1 9 - 10 1227 112
GenBank mRNA AK094247.1 - 6 - 7 800 112
AK127702.1 - 12 - 13 2030 112
BC064367.1 - 10 - 11 1430 112
CR749467.1 - 13 - 14 2114 112

View Full Product Details