Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLIS2
Entrez Gene ID
84662
Gene Name
GLIS family zinc finger 2
Gene Aliases
NKL, NPHP7
Chromosome Location
Chr.16: 4314761 - 4339597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318918.1 NP_001305847.1 6 - 7 1478 55
NM_032575.2 NP_115964.2 4 - 5 714 55
XM_005255641.4 XP_005255698.1 5 - 6 779 55
GenBank mRNA AF325914.1 - 4 - 5 714 55
AK095566.1 - 1 - 2 54 55
AK126918.1 - 6 - 7 1478 55

View Full Product Details