The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152342.3 | NP_689555.2 | 2 - 3 | 725 | 88 |
XM_011522866.1 | XP_011521168.1 | 2 - 3 | 737 | 88 | |
XM_011522867.2 | XP_011521169.1 | 2 - 3 | 1280 | 88 | |
GenBank mRNA | AK096185.1 | - | 2 - 3 | 725 | 88 |
AY273798.1 | - | 2 - 3 | 765 | 88 | |
BC069440.1 | - | 2 - 3 | 670 | 88 | |
BC100803.1 | - | 2 - 3 | 644 | 88 | |
BC100804.1 | - | 2 - 3 | 644 | 88 | |
BC100805.1 | - | 2 - 3 | 644 | 88 | |
BC100806.1 | - | 2 - 3 | 644 | 88 |