Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDYL2
Entrez Gene ID
124359
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
PCCP1
Chromosome Location
Chr.16: 80597899 - 80805043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152342.3 NP_689555.2 3 - 4 939 60
XM_011522866.1 XP_011521168.1 3 - 4 951 60
XM_011522867.2 XP_011521169.1 3 - 4 1494 60
GenBank mRNA AK096185.1 - 3 - 4 939 60
AX781670.1 - 3 - 4 929 60
AX781671.1 - 3 - 4 837 60
AY273798.1 - 3 - 4 979 60
BC069440.1 - 3 - 4 884 60
BC100803.1 - 3 - 4 858 60
BC100804.1 - 3 - 4 858 60
BC100805.1 - 3 - 4 858 60
BC100806.1 - 3 - 4 858 60

View Full Product Details