The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122608.2 | NP_001116080.1 | 6 - 7 | 821 | 80 |
NM_001171991.2 | NP_001165462.1 | 6 - 7 | 784 | 80 | |
NM_005333.4 | NP_005324.3 | 6 - 7 | 879 | 80 | |
GenBank mRNA | AK097815.1 | - | 6 - 7 | 806 | 80 |
BC001691.2 | - | 6 - 7 | 728 | 80 | |
BC095455.1 | - | 6 - 7 | 822 | 80 | |
CR749578.1 | - | 6 - 7 | 836 | 80 | |
U36787.1 | - | 6 - 7 | 803 | 80 |