The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015191.2 | NP_056006.1 | 1 - 2 | 308 | 64 |
GenBank mRNA | AB018324.3 | - | 1 - 2 | 209 | 64 |
AB084424.1 | - | 1 - 2 | 129 | 64 | |
AB096248.1 | - | 1 - 2 | 214 | 64 | |
AK000396.1 | - | 1 - 2 | 214 | 64 | |
AK291233.1 | - | 1 - 2 | 243 | 64 | |
AL136764.2 | - | 1 - 2 | 231 | 64 | |
BC113459.1 | - | 1 - 2 | 165 | 64 | |
BC117183.1 | - | 1 - 2 | 165 | 64 | |
BM799630.1 | - | 1 - 2 | 262 | 64 | |
DA333935.1 | - | 1 - 2 | 308 | 64 |