Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRBP2
Entrez Gene ID
340371
Gene Name
nuclear receptor binding protein 2
Gene Aliases
TRG16, pp9320
Chromosome Location
Chr.8: 143829776 - 143840974 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178564.3 NP_848659.2 14 - 15 1408 66
XM_017013378.1 XP_016868867.1 14 - 15 1408 66
XM_017013379.1 XP_016868868.1 14 - 15 1408 66
XM_017013380.1 XP_016868869.1 14 - 15 1408 66
XM_017013381.1 XP_016868870.1 14 - 15 1408 66
XM_017013382.1 XP_016868871.1 14 - 15 1408 66
XM_017013383.1 XP_016868872.1 14 - 15 1408 66
GenBank mRNA AF318376.1 - 4 - 5 748 66
AK094017.1 - 1 - 2 117 66
AK308967.1 - 1 - 2 99 66
AL137662.1 - 12 - 13 1337 66
AL834530.1 - 7 - 8 647 66
AY277601.1 - 4 - 5 735 66
BC071605.1 - 13 - 14 1571 66
BC113873.1 - 11 - 12 1412 66
BC114481.1 - 12 - 13 1309 66
CR457350.1 - 6 - 7 539 66

View Full Product Details