The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178088.1 | NP_001171559.1 | 22 - 23 | 3372 | 109 |
NM_003898.3 | NP_003889.1 | 23 - 24 | 3449 | 109 | |
XM_005267198.4 | XP_005267255.1 | 22 - 23 | 3380 | 109 | |
XM_005267199.2 | XP_005267256.1 | 22 - 23 | 3253 | 109 | |
XM_005267200.1 | XP_005267257.1 | 12 - 13 | 2147 | 109 | |
XM_006715592.3 | XP_006715655.1 | 23 - 24 | 3449 | 109 | |
XM_011536225.1 | XP_011534527.1 | 23 - 24 | 3489 | 109 | |
XM_011536226.2 | XP_011534528.1 | 17 - 18 | 4353 | 109 | |
XM_011536227.2 | XP_011534529.1 | 23 - 24 | 3449 | 109 | |
XM_011536228.2 | XP_011534530.1 | 17 - 18 | 4410 | 109 | |
XM_017011406.1 | XP_016866895.1 | 22 - 23 | 3437 | 109 | |
GenBank mRNA | AB002346.2 | - | 22 - 23 | 3185 | 109 |
AF039945.1 | - | 22 - 23 | 3185 | 109 | |
AF318616.1 | - | 23 - 24 | 3426 | 109 | |
AK074294.1 | - | 6 - 7 | 2665 | 109 | |
AK296935.1 | - | 10 - 11 | 1554 | 109 | |
AL157424.1 | - | 17 - 18 | 2828 | 110 | |
AY152396.1 | - | 23 - 24 | 3351 | 109 | |
BC043277.2 | - | 10 - 11 | 1782 | 109 | |
BX648749.1 | - | 22 - 23 | 3279 | 109 |