The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014159.6 | NP_054878.5 | 20 - 21 | 7587 | 113 |
XM_011533631.2 | XP_011531933.1 | 21 - 22 | 9083 | 113 | |
XM_011533632.2 | XP_011531934.1 | 20 - 21 | 7659 | 113 | |
XM_011533633.2 | XP_011531935.1 | 20 - 21 | 8950 | 113 | |
XM_011533634.1 | XP_011531936.1 | 20 - 21 | 7612 | 113 | |
XM_017006270.1 | XP_016861759.1 | 18 - 19 | 8786 | 113 | |
GenBank mRNA | AB051519.2 | - | 18 - 19 | 5587 | 113 |
AF049103.1 | - | 4 - 5 | 377 | 113 | |
AJ238403.1 | - | 18 - 19 | 6434 | 113 | |
AK026837.1 | - | 6 - 7 | 1245 | 113 | |
AK095494.1 | - | 13 - 14 | 2043 | 113 | |
AL713692.1 | - | 6 - 7 | 853 | 113 | |
AY576987.1 | - | 18 - 19 | 6096 | 113 | |
AY576988.1 | - | 20 - 21 | 6175 | 113 | |
BC090954.1 | - | 18 - 19 | 5377 | 113 | |
BC117162.1 | - | 18 - 19 | 6210 | 113 | |
BC117164.1 | - | 18 - 19 | 6210 | 113 | |
BX649110.1 | - | 18 - 19 | 7354 | 113 |