Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
Chromosome Location
Chr.12: 8822472 - 8887202 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144670.5 NP_653271.2 1 - 2 246 59
XM_011520566.2 XP_011518868.1 1 - 2 164 59
XM_011520567.2 XP_011518869.1 1 - 2 164 59
XM_017018868.1 XP_016874357.1 1 - 2 164 59
XM_017018869.1 XP_016874358.1 1 - 2 164 59
XM_017018870.1 XP_016874359.1 1 - 2 164 59
GenBank mRNA AK122624.1 - 1 - 2 140 59
AK290355.1 - 1 - 2 99 59
AL832139.1 - 1 - 2 97 59
BX648231.1 - 1 - 2 98 59
DA436610.1 - 1 - 2 164 59
DA456454.1 - 1 - 2 99 59
DA461408.1 - 1 - 2 246 59

View Full Product Details