Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPD4
Entrez Gene ID
167153
Gene Name
PAP associated domain containing 4
Gene Aliases
GLD2, TUT2
Chromosome Location
Chr.5: 79612416 - 79686648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114393.1 NP_001107865.1 12 - 13 1487 94
NM_001114394.1 NP_001107866.1 11 - 12 1762 94
NM_001297744.1 NP_001284673.1 12 - 13 1475 94
NM_001297745.1 NP_001284674.1 10 - 11 1633 94
NM_173797.3 NP_776158.2 12 - 13 1521 94
XM_011543219.2 XP_011541521.1 11 - 12 1841 94
XM_011543220.2 XP_011541522.1 12 - 13 1911 94
XM_011543221.2 XP_011541523.1 13 - 14 2000 94
XM_011543222.2 XP_011541524.1 12 - 13 1930 94
XM_011543223.2 XP_011541525.1 12 - 13 1550 94
XM_011543224.2 XP_011541526.1 12 - 13 1549 94
XM_011543225.2 XP_011541527.1 13 - 14 1511 94
XM_011543226.2 XP_011541528.1 13 - 14 1631 94
XM_011543227.2 XP_011541529.1 14 - 15 1720 94
XM_011543228.2 XP_011541530.1 11 - 12 1829 94
XM_011543229.2 XP_011541531.1 10 - 11 1712 94
XM_011543232.2 XP_011541534.1 7 - 8 627 94
XM_011543233.2 XP_011541535.1 7 - 8 615 94
XM_017009151.1 XP_016864640.1 13 - 14 1988 94
XM_017009152.1 XP_016864641.1 12 - 13 1899 94
XM_017009153.1 XP_016864642.1 12 - 13 1836 94
XM_017009154.1 XP_016864643.1 13 - 14 1913 94
XM_017009155.1 XP_016864644.1 12 - 13 1855 94
XM_017009156.1 XP_016864645.1 13 - 14 2145 94
XM_017009157.1 XP_016864646.1 11 - 12 1754 94
XM_017009158.1 XP_016864647.1 12 - 13 1824 94
XM_017009159.1 XP_016864648.1 12 - 13 1843 94
XM_017009160.1 XP_016864649.1 13 - 14 1424 94
XM_017009161.1 XP_016864650.1 13 - 14 1918 94
XM_017009162.1 XP_016864651.1 13 - 14 1494 94
GenBank mRNA AK095818.1 - 12 - 13 1368 94
AL833136.1 - 10 - 11 1619 94
BC026061.1 - 13 - 14 1520 94
BC047581.1 - 12 - 13 1454 94

View Full Product Details