The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152334.2 | NP_689547.2 | 3 - 4 | 623 | 72 |
XM_006720386.3 | XP_006720449.1 | 3 - 4 | 785 | 72 | |
XM_017021912.1 | XP_016877401.1 | 3 - 4 | 785 | 72 | |
GenBank mRNA | AK056653.1 | - | 3 - 4 | 613 | 72 |
AL833188.1 | - | 3 - 4 | 289 | 72 | |
BC132671.1 | - | 3 - 4 | 591 | 72 | |
BC136315.1 | - | 3 - 4 | 591 | 72 | |
BC143943.1 | - | 3 - 4 | 591 | 72 |