Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARM1
Entrez Gene ID
10498
Gene Name
coactivator associated arginine methyltransferase 1
Gene Aliases
PRMT4
Chromosome Location
Chr.19: 10871577 - 10923078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199141.1 NP_954592.1 7 - 8 1067 109
XM_005259708.4 XP_005259765.1 7 - 8 1033 109
XM_011527638.2 XP_011525940.1 6 - 7 823 109
GenBank mRNA AF055027.1 - 2 - 3 242 109
AL833242.1 - 7 - 8 816 109
BC046240.1 - - 364 109

View Full Product Details