The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099409.1 | NP_001092879.1 | 12 - 13 | 4199 | 112 |
XM_005273867.3 | XP_005273924.1 | 14 - 15 | 2612 | 112 | |
XM_005273868.3 | XP_005273925.1 | 13 - 14 | 2437 | 112 | |
XM_005273869.3 | XP_005273926.1 | 13 - 14 | 2139 | 112 | |
XM_005273870.3 | XP_005273927.1 | 12 - 13 | 1964 | 112 | |
XM_006718486.3 | XP_006718549.1 | 13 - 14 | 4236 | 112 | |
XM_011544886.2 | XP_011543188.1 | 5 - 6 | 996 | 112 | |
XM_017017467.1 | XP_016872956.1 | 13 - 14 | 3986 | 112 | |
XM_017017468.1 | XP_016872957.1 | 13 - 14 | 3963 | 112 | |
XM_017017469.1 | XP_016872958.1 | 11 - 12 | 1927 | 112 | |
GenBank mRNA | AK024451.1 | - | 8 - 9 | 3611 | 112 |
AL834433.1 | - | 12 - 13 | 3896 | 112 | |
BC024207.2 | - | 1 - 2 | 96 | 112 |