The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040202.1 | NP_001035292.1 | 3 - 4 | 712 | 80 |
XM_005262769.4 | XP_005262826.1 | 3 - 4 | 712 | 80 | |
XM_017007785.1 | XP_016863274.1 | 3 - 4 | 712 | 80 | |
GenBank mRNA | AK055774.1 | - | 3 - 4 | 500 | 80 |
AK291232.1 | - | 3 - 4 | 692 | 80 | |
AX775925.1 | - | 3 - 4 | 706 | 80 | |
AY424281.1 | - | 3 - 4 | 639 | 80 | |
BC029604.1 | - | 2 - 3 | 480 | 80 | |
BC031256.1 | - | 3 - 4 | 678 | 80 | |
BC047510.1 | - | 3 - 4 | 687 | 80 | |
BX648346.1 | - | 2 - 3 | 343 | 80 |