Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
Chromosome Location
Chr.17: 10697610 - 10711558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005256738.2 XP_005256795.1 3 - 4 791 68
GenBank mRNA BC001294.2 - 2 - 3 834 68

View Full Product Details