Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHD
Entrez Gene ID
6007
Gene Name
Rh blood group D antigen
Gene Aliases
CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
Chromosome Location
Chr.1: 25272393 - 25330445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127691.2 NP_001121163.1 2 - 3 394 92
NM_001282867.1 NP_001269796.1 2 - 3 394 92
NM_001282868.1 NP_001269797.1 2 - 3 394 92
NM_001282869.1 NP_001269798.1 2 - 3 394 92
NM_001282870.1 NP_001269799.1 2 - 3 394 92
NM_001282871.1 NP_001269800.1 2 - 3 394 92
NM_001282872.1 NP_001269801.1 2 - 3 394 92
NM_016124.4 NP_057208.2 2 - 3 394 92
XM_017002015.1 XP_016857504.1 2 - 3 487 92
GenBank mRNA AB018966.1 - 2 - 3 332 92
AB018967.1 - 2 - 3 332 92
AB018968.1 - 2 - 3 332 92
AB018969.1 - 2 - 3 332 92
AB037270.1 - 2 - 3 332 92
AB046420.1 - 2 - 3 332 92
AF037626.1 - 2 - 3 332 92
AF177938.1 - 2 - 3 332 92
AF187846.1 - 2 - 3 332 92
AF510069.1 - 2 - 3 375 92
AF510070.1 - 2 - 3 375 92
AJ704215.2 - 2 - 3 332 92
AK297151.1 - 2 - 3 487 92
AM183924.3 - 2 - 3 332 92
AY449381.1 - 2 - 3 332 92
AY449382.1 - 2 - 3 332 92
AY449383.1 - 2 - 3 332 92
AY449384.1 - 2 - 3 332 92
AY449385.1 - 2 - 3 332 92
AY751491.1 - 2 - 3 332 92
AY751492.1 - 2 - 3 332 92
AY751493.1 - 2 - 3 332 92
AY751494.1 - 2 - 3 332 92
AY751495.1 - 2 - 3 332 92
AY751496.1 - 2 - 3 332 92
BC139922.1 - 2 - 3 382 92
DQ451878.1 - 2 - 3 351 92
EU557240.1 - 2 - 3 335 92
EU557241.1 - 2 - 3 335 92
EU661905.1 - 2 - 3 272 92
EU661906.1 - 2 - 3 272 92
FR745438.1 - 2 - 3 332 92
FR745439.1 - 2 - 3 332 92
FR745440.1 - 2 - 3 332 92
FR745441.1 - 2 - 3 332 92
HE657775.1 - 2 - 3 332 92
JF436967.1 - 2 - 3 332 92
JN635688.1 - 2 - 3 332 92
JN635689.1 - 2 - 3 332 92
JQ065733.1 - 2 - 3 332 92
JQ424879.1 - 2 - 3 390 92
KT240043.1 - 2 - 3 332 92
L08429.1 - 2 - 3 332 92
LC053447.1 - 2 - 3 332 92
LN714468.1 - 2 - 3 332 92
LN824132.1 - 2 - 3 332 92
S57971.1 - 2 - 3 332 92
S70174.1 - 2 - 3 332 92
S73913.1 - 2 - 3 332 92
S78509.1 - 2 - 3 332 92
X63094.1 - 2 - 3 390 92
X63097.1 - 2 - 3 348 92

Gene Symbol
RHCE
Entrez Gene ID
6006
Gene Name
Rh blood group CcEe antigens
Gene Aliases
CD240CE, RH, RH30A, RHC, RHE, RHIXB, RHPI, Rh4, RhIVb(J), RhVI, RhVIII
Chromosome Location
Chr.1: 25362249 - 25430193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB030388.1 - 2 - 3 332 92
AB049753.1 - 2 - 3 332 92
JF436968.1 - 2 - 3 332 92
KF515558.1 - 2 - 3 332 92

View Full Product Details