The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099337.1 | NP_001092807.1 | 2 - 3 | 294 | 77 |
NM_144576.3 | NP_653177.3 | 2 - 3 | 538 | 77 | |
GenBank mRNA | AK057003.1 | - | 2 - 3 | 454 | 77 |
AK057014.1 | - | 1 - 2 | 255 | 77 | |
AY358861.1 | - | 2 - 3 | 226 | 77 | |
BC002435.2 | - | 1 - 2 | 115 | 77 | |
BC026922.1 | - | 2 - 3 | 294 | 77 | |
BC073923.1 | - | 2 - 3 | 422 | 77 |