Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM243
Entrez Gene ID
79161
Gene Name
transmembrane protein 243
Gene Aliases
C7orf23, MM-TRAG, MMTRAG
Chromosome Location
Chr.7: 87195593 - 87220603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005250586.3 XP_005250643.1 2 - 3 507 134
XM_005250587.4 XP_005250644.1 2 - 3 641 134
XM_005250588.3 XP_005250645.1 2 - 3 339 134
XM_005250589.3 XP_005250646.1 2 - 3 507 134
XM_017012616.1 XP_016868105.1 1 - 2 928 134
XM_017012617.1 XP_016868106.1 2 - 3 630 134
XM_017012618.1 XP_016868107.1 2 - 3 374 134
GenBank mRNA AK313794.1 - 1 - 2 309 134
BC002837.2 - 1 - 2 291 134
BT006973.1 - 1 - 2 79 134
CB993361.1 - 2 - 3 548 134

View Full Product Details